Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014577.4 | XP_004014626.1 | 11 - 12 | 1150 | 58 | |
XM_027977208.1 | XP_027833009.1 | 10 - 11 | 840 | 58 | ||
XM_027977209.1 | XP_027833010.1 | 10 - 11 | 837 | 58 | ||
XM_027977210.1 | XP_027833011.1 | 10 - 11 | 840 | 58 | ||
XM_027977211.1 | XP_027833012.1 | 10 - 11 | 840 | 58 | ||
XM_027977213.1 | XP_027833014.1 | 10 - 11 | 840 | 58 | ||
XM_027977214.1 | XP_027833015.1 | 10 - 11 | 1003 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |