Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973995.1 | XP_027829796.1 | 6 - 7 | 940 | 61 | |
XM_027973996.1 | XP_027829797.1 | 4 - 5 | 1611 | 61 | ||
XM_027973997.1 | XP_027829798.1 | 6 - 7 | 664 | 61 |