Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015097916.2 | XP_014953402.2 | 3 - 4 | 801 | 83 | |
XM_027973202.1 | XP_027829003.1 | 3 - 4 | 699 | 83 |