Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006958.4 | XP_004007007.4 | 1 - 2 | 368 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |