Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010751.3 | XP_004010800.1 | 3 - 4 | 423 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |