Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962235.1 | XP_027818036.1 | 3 - 4 | 579 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |