Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098961.2 | XP_014954447.2 | 2 - 3 | 367 | 118 | |
XM_027974059.1 | XP_027829860.1 | 2 - 3 | 372 | 211 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |