Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC45A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972827.1 | XP_027828628.1 | 3 - 4 | 1094 | 91 | |
XM_027972829.1 | XP_027828630.1 | 3 - 4 | 1092 | 91 | ||
XM_027972830.1 | XP_027828631.1 | 3 - 4 | 1096 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |