Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METRNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098887.2 | XP_014954373.2 | 3 - 4 | 811 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |