Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAB21L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973497.1 | XP_027829298.1 | 4 - 5 | 375 | 78 | |
XM_027973498.1 | XP_027829299.1 | 4 - 5 | 391 | 78 |