Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104301.2 | XP_014959787.2 | 2 - 3 | 591 | 102 | |
XM_027962298.1 | XP_027818099.1 | 2 - 3 | 593 | 102 | ||
XM_027962299.1 | XP_027818100.1 | 2 - 3 | 593 | 102 | ||
XM_027962300.1 | XP_027818101.1 | 2 - 3 | 593 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |