Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104301.2 | XP_014959787.2 | 3 - 4 | 754 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |