Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973435.1 | XP_027829236.1 | 6 - 7 | 556 | 91 | |
XM_027973436.1 | XP_027829237.1 | 6 - 7 | 548 | 91 | ||
XM_027973437.1 | XP_027829238.1 | 5 - 6 | 323 | 91 | ||
XM_027973438.1 | XP_027829239.1 | 6 - 7 | 525 | 91 | ||
XM_027973439.1 | XP_027829240.1 | 6 - 7 | 524 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |