Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102776.2 | XP_014958262.2 | 1 - 2 | 500 | 59 | |
XM_027958773.1 | XP_027814574.1 | 2 - 3 | 336 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |