Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008025.4 | XP_004008074.3 | 2 - 3 | 1766 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |