Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970152.1 | XP_027825953.1 | 7 - 8 | 2633 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |