Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012179414.3 | XP_012034804.2 | 4 - 5 | 788 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |