Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979116.1 | XP_027834917.1 | 4 - 5 | 554 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |