Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002146.4 | XP_004002195.3 | 1 - 2 | 705 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |