Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004497.4 | XP_004004546.1 | 5 - 6 | 839 | 76 | |
XM_012130961.3 | XP_011986351.1 | 5 - 6 | 1088 | 76 | ||
XM_012130979.3 | XP_011986369.1 | 5 - 6 | 950 | 76 | ||
XM_012131005.3 | XP_011986395.1 | 5 - 6 | 966 | 76 | ||
XM_012131015.3 | XP_011986405.1 | 5 - 6 | 744 | 76 | ||
XM_027964710.1 | XP_027820511.1 | 5 - 6 | 1493 | 76 | ||
XM_027964711.1 | XP_027820512.1 | 5 - 6 | 1104 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |