Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004009400.4 | XP_004009449.2 | 2 - 3 | 532 | 68 | |
XM_012178313.3 | XP_012033703.2 | 2 - 3 | 535 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |