Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013031.3 | XP_004013080.2 | 5 - 6 | 1369 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |