Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101113346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975299.1 | XP_027831100.1 | 3 - 4 | 1448 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |