Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977022.1 | XP_027832823.1 | 6 - 7 | 2842 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |