Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977607.1 | XP_027833408.1 | 3 - 4 | 448 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |