Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004503.4 | XP_004004552.1 | 2 - 3 | 315 | 82 | |
XM_004004505.4 | XP_004004554.1 | 2 - 3 | 242 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |