Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018995.4 | XP_004019044.2 | 3 - 4 | 1262 | 127 | |
XM_027958906.1 | XP_027814707.1 | 3 - 4 | 1115 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |