Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964139.1 | XP_027819940.1 | 4 - 5 | 1542 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |