Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006523.2 | XP_004006572.1 | 12 - 13 | 1631 | 102 | |
XM_012174649.2 | XP_012030039.1 | 12 - 13 | 1641 | 102 | ||
XM_012174650.2 | XP_012030040.1 | 11 - 12 | 1536 | 102 | ||
XM_012174651.2 | XP_012030041.1 | 11 - 12 | 1527 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |