Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRGPRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959556.1 | XP_027815357.1 | 2 - 3 | 301 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Ovis aries |