Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APEX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106627.3 | XP_011962017.1 | 3 - 4 | 351 | 93 | |
XM_027963026.1 | XP_027818827.1 | 3 - 4 | 554 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |