Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103001.3 | XP_011958391.3 | 2 - 3 | 1192 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |