Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096165.2 | XP_014951651.2 | 1 - 2 | 448 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |