Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006200.4 | XP_004006249.1 | 3 - 4 | 628 | 65 | |
XM_027967152.1 | XP_027822953.1 | 3 - 4 | 628 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |