Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963519.1 | XP_027819320.1 | 5 - 6 | 1574 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |