Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017018.4 | XP_004017067.1 | 2 - 3 | 1162 | 82 | |
XM_012097000.3 | XP_011952390.1 | 2 - 3 | 1160 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |