Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R14D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010454.4 | XP_004010503.1 | 3 - 4 | 552 | 119 | |
XM_015096950.2 | XP_014952436.1 | 5 - 6 | 502 | 119 | ||
XM_015096951.2 | XP_014952437.1 | 3 - 4 | 459 | 119 | ||
XM_015096952.2 | XP_014952438.2 | 3 - 4 | 561 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |