Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979417.1 | XP_027835218.1 | 6 - 7 | 1473 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |