Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101115110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006861.4 | XP_004006910.2 | 1 | 373 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |