Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETHE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978406.1 | XP_027834207.1 | 6 - 7 | 784 | 71 | |
XM_027978407.1 | XP_027834208.1 | 5 - 6 | 592 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |