Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDXACB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016016.4 | XP_004016065.2 | 2 - 3 | 354 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |