Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC16A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098988.2 | XP_014954474.2 | 4 - 5 | 667 | 88 | |
XM_027973993.1 | XP_027829794.1 | 3 - 4 | 1481 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |