Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012175453.2 | XP_012030843.2 | 4 - 5 | 579 | 98 | |
XM_012175454.3 | XP_012030844.2 | 4 - 5 | 459 | 98 | ||
XM_012175455.3 | XP_012030845.2 | 4 - 5 | 1381 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |