Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013832.4 | XP_004013881.1 | 2 - 3 | 985 | 105 | |
XM_015099338.2 | XP_014954824.1 | 2 - 3 | 579 | 105 | ||
XM_015099339.2 | XP_014954825.1 | 2 - 3 | 1935 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |