Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964045.1 | XP_027819846.1 | 4 - 5 | 546 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |