Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012103989.2 | XP_011959379.2 | 4 - 5 | 437 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |