Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962577.1 | XP_027818378.1 | 7 - 8 | 9205 | 89 | |
XM_027962578.1 | XP_027818379.1 | 8 - 9 | 9074 | 89 | ||
XM_027962579.1 | XP_027818380.1 | 6 - 7 | 8456 | 89 |