Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMILIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103646.2 | XP_014959132.2 | 7 - 8 | 3204 | 68 | |
XM_027960820.1 | XP_027816621.1 | 6 - 7 | 2995 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |