Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094306.2 | XP_014949792.2 | 3 - 4 | 603 | 107 | |
XM_015094307.2 | XP_014949793.2 | 2 - 3 | 518 | 107 | ||
XM_015094309.2 | XP_014949795.2 | 3 - 4 | 603 | 107 | ||
XM_015094311.2 | XP_014949797.2 | 3 - 4 | 603 | 107 | ||
XM_027966692.1 | XP_027822493.1 | 3 - 4 | 603 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |