Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101115856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022132.4 | XP_004022181.1 | 2 - 3 | 1901 | 69 |